tert-Butyl-4-((tosyloxy)methyl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-(tosyloxymethyl)piperidine, 1-(tert-Butoxycarbonyl)-4-(tosyloxymethyl)piperidine, tert-Butyl 4-((tosyloxy)methyl)piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-[(p-toluenesulphonyloxy)methyl]piperidine

A336184-10G A336184-250MG A336184-1G A336184-25G A336184-5G
103825-710EA 6 USD
103825-710 103836-686 76724-248 76724-250 76839-688 76839-690 76839-692
tert-Butyl-4-((tosyloxy)methyl)piperidine-1-carboxylate 97%
tert-Butyl-4-((tosyloxy)methyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₇NO₅S
MW: 369.48 g/mol
Boiling Pt: 487.7 °C (760 mm Hg)
Flash Pt: 248.803 °C
Storage Temperature: Ambient
MDL Number: MFCD05864740
CAS Number: 166815-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR