tert-Butyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-(methanesulphonyloxy)piperidine, tert-Butyl 4-((methylsulphonyl)oxy)piperidine-1-carboxylate, tert-Butyl 4-(Methanesulphonyloxy)piperidine-1-carboxylate, 4-(Methanesulphonyloxy)piperidine-1-carboxylic acid tert-Butyl ester, 1-(tert-Butoxycarbonyl)-4-(methanesulphonyloxy)piperidine, 1-(tert-Butoxycarbonyl)-4-piperidyl Methanesulphonate

A456840-100G A456840-10G A456840-500G A456840-1G A456840-25G A456840-5G
103827-862EA 5 USD
103827-862 103838-938 76804-578 76804-580 76804-582 76842-794 76842-796 76842-798
tert-Butyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate 98%
tert-Butyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₅S
MW: 279.3500 g/mol
Melting Pt: 83…87 °C
Flash Pt: 200.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04116220
CAS Number: 141699-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR