tert-Butyl-3-oxocyclobutylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: Carbamic acid, N-(3-oxocyclobutyl)-, 1,1-dimethylethyl ester, 2-Methyl-2-propanyl (3-oxocyclobutyl)carbamate, tert-Butyl 3-oxocyclobutylcarbamate

A264872-100G A264872-250MG A264872-1G A264872-25G A264872-5G A264872-10G
103812-702EA 16 USD
103812-702 103852-596 76692-756 76692-758 77092-256 77092-258 77092-260 77568-518
tert-Butyl-3-oxocyclobutylcarbamate 97%
tert-Butyl-3-oxocyclobutylcarbamate

Precaución: For research use only.

Formula: C₉H₁₅NO₃
Boiling Pt: 302.1 °C (760 mm Hg)
Melting Pt: 120…125 °C
Flash Pt: 136.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09751872
CAS Number: 154748-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR