tert-Butyl-3-oxocyclobutanecarboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 3-oxocyclobutanecarboxylate
A251736-10G
A251736-1G
A251736-5G
A251736-100G
103812-004EA
6
USD
103812-004
103851-844
76786-828
77091-232
77091-234
77601-030
tert-Butyl-3-oxocyclobutanecarboxylate 95%
tert-Butyl-3-oxocyclobutanecarboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₄O₃ MW: 170.208 g/mol Boiling Pt: 230.4 °C (760 mm Hg) Flash Pt: 93.4 °C Storage Temperature: Freezer |
MDL Number:
MFCD12827541 CAS Number: 145549-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...