tert-Butyl-3-oxoazetidine-1-carboxylate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
N-Boc-3-azetidinone, Tert-butyl 3-oxoazetidine-1-carboxylate, tert-Butyl 3-oxoazetidine-1-carboxylate, 1-BOC-3-Azetidinone
A216673-25G
A216673-5G
A216673-100G
A216673-10G
A216673-500G
103849-292EA
15
USD
103849-292
77087-882
77087-884
77165-112
77165-114
77165-116
tert-Butyl-3-oxoazetidine-1-carboxylate 98%
tert-Butyl-3-oxoazetidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₁₃NO₃ MW: 171.2 g/mol Boiling Pt: 251.3 °C (760 mm Hg) Melting Pt: 49…52 °C Flash Pt: 101.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01861741 CAS Number: 398489-26-4 UN: 3335 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...