9-Boc-3-oxo-9-azabicyclo[3.3.1]nonane 97%

Supplier: Ambeed
Warning

A359818-1G A359818-25G A359818-5G A359818-100G A359818-250MG
103826-236EA 6 USD
103826-236 103837-244 76840-430 76840-432 76840-434 77106-898 77106-900
9-Boc-3-oxo-9-azabicyclo[3.3.1]nonane 97%
9-Boc-3-oxo-9-azabicyclo[3.3.1]nonane

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₃
Boiling Pt: 342.9 °C (760 mm Hg)
Flash Pt: 161.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09997838
CAS Number: 512822-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR