1-Boc-3-methylpiperazine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-methylpiperazine-1-carboxylate, 3-Methylpiperazine, N1-BOC protected

A143530-100G A143530-1G A143530-5G A143530-10G
103803-294EA 5 USD
103803-294 76689-088 77299-344 77316-570 77879-006
1-Boc-3-methylpiperazine 97%
1-Boc-3-methylpiperazine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
Boiling Pt: 268.7 °C (760 mm Hg)
Flash Pt: 116.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03001706
CAS Number: 120737-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR