tert-Butyl-3-methyleneazetidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methylideneazetidine, N-BOC protected, tert-Butyl 3-methyleneazetidine-1-carboxylate

A392854-25G A392854-1G A392854-5G A392854-10G
103826-910EA 12 USD
103826-910 103837-948 76876-802 77307-020 77307-022 77568-802
tert-Butyl-3-methyleneazetidine-1-carboxylate 97%
tert-Butyl-3-methyleneazetidine-1-carboxylate

Precaución: For research use only.

Formula: C₉H₁₅NO₂
MW: 169.23 g/mol
Boiling Pt: 214.8±29.0 °C (760 mm Hg)
Flash Pt: 83.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12031229
CAS Number: 934664-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR