tert-Butyl-3-iodoazetidine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A349320-1G
A349320-25G
A349320-5G
A349320-100G
A349320-10G
A349320-500G
103826-014EA
5
USD
103826-014
103837-004
76840-090
76840-092
76840-094
77106-762
77106-764
77685-990
tert-Butyl-3-iodoazetidine-1-carboxylate 97%
tert-Butyl-3-iodoazetidine-1-carboxylate
Formula:
C₈H₁₄INO₂ Boiling Pt: 282 °C (760 mm Hg) Flash Pt: 124.372 °C Storage Temperature: Ambient |
MDL Number:
MFCD09752821 CAS Number: 254454-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...