1-Boc-3-iodoindole 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-iodo-1H-indole-1-carboxylate

A288753-5G A288753-250MG
103813-808EA 34 USD
103813-808 103834-950 76837-516 77194-272
1-Boc-3-iodoindole 98%
1-Boc-3-iodoindole

Precaución: for Research Use Only

Formula: C₁₃H₁₄INO₂
MW: 343.16 g/mol
Boiling Pt: 388.372 °C (760 mm Hg)
Melting Pt: 36…40 °C
Flash Pt: 188.681 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05864781
CAS Number: 192189-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR