tert-Butyl-3-hydroxypent-4-enoate 97%

Supplier: Ambeed
Warning

A112335-10G A112335-1G A112335-5G A112335-25G A112335-100G
103798-000EA 8 USD
103798-000 103823-834 76869-798 77310-638 77310-640 77885-044 77885-043
tert-Butyl-3-hydroxypent-4-enoate 97%
tert-Butyl-3-hydroxypent-4-enoate

Precaución: for Research Use Only

Formula: C₉H₁₆O₃
Boiling Pt: 238.1±20.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD18207180
CAS Number: 122763-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR