tert-Butyl-3-hydroxyazetidine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-3-hydroxyazetidine, tert-Butyl 3-hydroxyazetidine-1-carboxylate, 1-BOC-3-Hydroxyazetidine, tert-butyl 3-hydroxyazetidine-1-carboxylate

A189343-25G A189343-5G A189343-100G A189343-10G A189343-1G A189343-500G
103846-772EA 6 USD
103846-772 77084-542 77084-544 77191-922 77191-924 77191-926 77191-928
tert-Butyl-3-hydroxyazetidine-1-carboxylate 98%
tert-Butyl-3-hydroxyazetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
Melting Pt: 36…43 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD04115305
CAS Number: 141699-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR