3-Hydroxy-1-oxa-8-azaspiro[4.5]decane-8-carboxylic acid tert-butyl ester 97%

Supplier: Ambeed
Warning

A108616-100MG A108616-250MG
103797-372EA 39 USD
103797-372 103823-152 76717-968 76717-970
3-Hydroxy-1-oxa-8-azaspiro[4.5]decane-8-carboxylic acid tert-butyl ester 97%
3-Hydroxy-1-oxa-8-azaspiro[4.5]decane-8-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
Boiling Pt: 391 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09749842
CAS Number: 240401-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR