1-Boc-azetidine-3-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3-Formylazetidine-1-carboxylic acid tert-Butyl ester, 1-(tert-Butoxycarbonyl)azetidine-3-carboxaldehyde, tert-Butyl 3-Formylazetidine-1-carboxylate, 1-Boc-azetidine-3-carboxaldehyde

A348795-1G A348795-25G A348795-5G A348795-10G A348795-250MG
103799-616EA 20 USD
103799-616 103825-998 103836-988 76840-066 76840-068 76840-070 76989-584 76989-586
1-Boc-azetidine-3-carboxaldehyde 97%
1-Boc-azetidine-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Boiling Pt: 256.6 °C (760 mm Hg)
Flash Pt: 109 °C
Storage Temperature: Freezer
MDL Number: MFCD06656142
CAS Number: 177947-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR