3-Fluoro-4-hydroxy-piperidine-1-carboxylic acid tert-Butyl ester 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-fluoro-4-hydroxypiperidine-1-carboxylate

A160297-100MG A160297-10G A160297-250MG A160297-1G A160297-5G
103805-360EA 47 USD
103805-360 103843-588 76973-544 76973-546 76973-548 77319-508 77319-510
3-Fluoro-4-hydroxy-piperidine-1-carboxylic acid tert-Butyl ester 97%
3-Fluoro-4-hydroxy-piperidine-1-carboxylic acid tert-Butyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₈FNO₃
MW: 219.253 g/mol
Boiling Pt: 300.8 °C (760 mm Hg)
Flash Pt: 135.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11977352
CAS Number: 373604-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR