1-(N-Boc)-3-cyanoazetidine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-cyanoazetidine-1-carboxylate, Azetidine-3-carbonitrile, N-BOC protected

A125916-100G A125916-10G A125916-250MG A125916-1G A125916-25G A125916-5G
103800-738EA 9 USD
103800-738 76996-104 76996-106 76996-108 77312-896 77312-898 77312-900
1-(N-Boc)-3-cyanoazetidine 97%
1-(N-Boc)-3-cyanoazetidine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₂
Flash Pt: 129.281 °C
Storage Temperature: Ambient
MDL Number: MFCD06796640
CAS Number: 142253-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR