tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Cyano-4-oxopyrrolidine-1-carboxylic acid tert-Butyl ester, tert-Butyl 3-cyano-4-oxopyrrolidine-1-carboxylate, 1-Boc-3-cyano-4-pyrrolidone, tert-Butyl 3-Cyano-4-oxopyrrolidine-1-carboxylate, 1-tert-Butoxycarbonyl-3-cyano-4-pyrrolidone, 1-Boc-3-cyano-4-pyrrolidinone, 1-tert-Butoxycarbonyl-3-cyano-4-pyrrolidinone
A212733-100G
A212733-10G
A212733-500G
A212733-1G
A212733-25G
A212733-5G
103818-232EA
9
USD
103818-232
103848-928
76770-588
76770-590
76770-592
77087-402
77303-250
77303-252
tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate 97%
tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₄N₂O₃ MW: 210.23 g/mol Boiling Pt: 360.7 °C (760 mm Hg) Melting Pt: 160…167 °C Flash Pt: 172 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01318456 CAS Number: 175463-32-8 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...