3-Cyano-4-oxopiperidine-1-carboxylic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-cyano-4-oxopiperidine-1-carboxylate

A103287-10G A103287-250MG A103287-1G
103796-524EA 14 USD
103796-524 103822-228 76780-616 76780-618 77283-074
3-Cyano-4-oxopiperidine-1-carboxylic acid tert-butyl ester 95%
3-Cyano-4-oxopiperidine-1-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₃
MW: 224.26 g/mol
Boiling Pt: 375.1 °C (760 mm Hg)
Melting Pt: 102…106 °C
Flash Pt: 180.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11109814
CAS Number: 914988-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR