tert-Butyl-3-bromopyrrolidine-1-carboxylate 98%
Supplier: Ambeed
|
Danger
|
A116101-250MG
A116101-5G
A116101-25G
A116101-10G
A116101-100G
103798-674EA
12
USD
103798-674
103824-538
76718-308
77297-420
77570-982
77599-672
77878-872
tert-Butyl-3-bromopyrrolidine-1-carboxylate 98%
tert-Butyl-3-bromopyrrolidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₆BrNO₂ Boiling Pt: 277.5±33.0 °C (760 mm Hg) Melting Pt: 47…52 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08752478 CAS Number: 939793-16-5 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...