1-Boc-3-bromoazetidine 97%

Supplier: Ambeed
Warning

A148938-100MG A148938-10G A148938-250MG A148938-1G A148938-25G A148938-5G A148938-100G
103803-986EA 9 USD
103803-986 103842-106 76812-108 76812-110 76812-112 77317-576 77317-578 77317-580 77883-178
1-Boc-3-bromoazetidine 97%
1-Boc-3-bromoazetidine

Precaución: for Research Use Only

Formula: C₈H₁₄BrNO₂
Boiling Pt: 258 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16658899
CAS Number: 1064194-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR