tert-Butyl 3-bromo-6,7-dihydropyrazolo[1,5-a]pyrazine-5(4H)-carboxylate 97%

Supplier: Ambeed
Warning

A246976-100MG A246976-10G A246976-250MG A246976-1G A246976-5G
103811-742EA 83 USD
103811-742 103851-576 76976-596 76976-598 76976-600 77090-878 77090-880
tert-Butyl 3-bromo-6,7-dihydropyrazolo[1,5-a]pyrazine-5(4H)-carboxylate 97%
tert-Butyl 3-bromo-6,7-dihydropyrazolo[1,5-a]pyrazine-5(4H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₆BrN₃O₂
Boiling Pt: 399.4 °C (760 mm Hg)
Flash Pt: 195.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13190301
CAS Number: 1196154-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR