tert-Butyl 3-bromo-5,6-dihydroimidazo[1,5-a]pyrazine-7(8H)-carboxylate 95%

Supplier: Ambeed
Warning

A265810-100MG A265810-250MG
103812-758EA 355 USD
103812-758 103852-654 77031-840 77031-842
tert-Butyl 3-bromo-5,6-dihydroimidazo[1,5-a]pyrazine-7(8H)-carboxylate 95%
tert-Butyl 3-bromo-5,6-dihydroimidazo[1,5-a]pyrazine-7(8H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₆BrN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD13152219
CAS Number: 1188264-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR