tert-Butyl-3-bromo-5,6-dihydro[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-carboxylate 97%

Supplier: Ambeed
Warning

A275016-1G A275016-100MG A275016-250MG A275016-5G
103813-236EA 63 USD
103813-236 103853-164 77093-004 77166-544 77166-546 77305-188
tert-Butyl-3-bromo-5,6-dihydro[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-carboxylate 97%
tert-Butyl-3-bromo-5,6-dihydro[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₅BrN₄O₂
Boiling Pt: 430.4 °C (760 mm Hg)
Flash Pt: 195.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08448169
CAS Number: 723286-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR