tert-Butyl-3-bromo-4-oxopiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A207023-100G A207023-10G A207023-1G A207023-5G A207023-25G
103817-754EA 10 USD
103817-754 103848-434 77012-826 77012-828 77086-698 77086-700 77303-018
tert-Butyl-3-bromo-4-oxopiperidine-1-carboxylate 95%
tert-Butyl-3-bromo-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆BrNO₃
Melting Pt: 194 °C
Flash Pt: 155.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06738530
CAS Number: 188869-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR