1-Boc-3-bromoindole 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-bromo-1H-indole-1-carboxylate, tert-Butyl-3-bromo-1H-indole-1-carboxylate, 3-Bromoindole-1-carboxylic acid tert-butyl ester

A114352-100G A114352-100MG A114352-10G A114352-250MG A114352-1G A114352-5G A114352-25G
103798-388EA 8 USD
103798-388 103824-228 77009-102 77009-104 77009-106 77009-108 77311-036 77311-038 77570-796
1-Boc-3-bromoindole 95%
1-Boc-3-bromoindole

Precaución: for Research Use Only

Formula: C₁₃H₁₄BrNO₂
MW: 296.16 g/mol
Boiling Pt: 367.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD05864772
CAS Number: 143259-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR