tert-Butyl 3-aminopropanoate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-Aminopropanoate

A154601-250MG A154601-1G A154601-100MG
103804-660EA 35 USD
103804-660 103842-850 76871-792 77318-582 77671-180
tert-Butyl 3-aminopropanoate 97%
tert-Butyl 3-aminopropanoate

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
Boiling Pt: 192.3 °C (760 mm Hg)
Flash Pt: 64 °C
Storage Temperature: Freezer
MDL Number: MFCD06212562
CAS Number: 15231-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR