3-Amino-N-(tert-butoxycarbonyl)benzylamine 97%

Supplier: Ambeed
Warning

Synonyms: alpha-(Boc-amino)-m-toluidine, 3-Amino-N-Boc-benzylamine, tert-Butyl (3-Aminobenzyl)carbamate, alpha-(tert-Butoxycarbonyl)amino-m-toluidine, 3-[(tert-Butoxycarbonylamino)methyl]aniline, 3-[(Boc-amino)methyl]aniline, (3-Aminobenzyl)carbamic Acid tert-Butyl, tert-Butyl 3-aminobenzylcarbamate

A184838-250MG A184838-1G A184838-25G
103815-792EA 43 USD
103815-792 103846-358 76784-706 77083-982 77871-921
3-Amino-N-(tert-butoxycarbonyl)benzylamine 97%
3-Amino-N-(tert-butoxycarbonyl)benzylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MW: 222.29 g/mol
Boiling Pt: 386 °C (760 mm Hg)
Flash Pt: 187.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04974010
CAS Number: 147291-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR