3-Amino-6,6-dimethyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole, N5-BOC protected 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-amino-6,6-dimethylpyrrolo[3,4-c]pyrazole-5(1H,4H,6H)-carboxylate, tert-Butyl 3-amino-4,6-dihydro-6,6-dimethylpyrrolo[3,4-c]pyrazole-5(1H)-carboxylate, 3-Amino-5-(tert-butoxycarbonyl)-6,6-dimethyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole

A150296-250MG A150296-1G A150296-5G
103804-142EA 20 USD
103804-142 103842-276 76783-354 77317-784 77317-786
3-Amino-6,6-dimethyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole, N5-BOC protected 95%
3-Amino-6,6-dimethyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole, N5-BOC protected

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₄O₂
MW: 252.313 g/mol
Boiling Pt: 420.6 °C (760 mm Hg)
Flash Pt: 208.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275051
CAS Number: 398491-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR