3-Boc-3,9-diazaspiro[5.5]undecane 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3,9-diazaspiro[5.5]undecane-3-carboxylate

A431618-25G A431618-5G A431618-100MG A431618-10G A431618-250MG A431618-100G
103827-524EA 34 USD
103827-524 103838-590 76842-282 76842-284 77279-056 77279-058 77279-060 77748-873
3-Boc-3,9-diazaspiro[5.5]undecane 97%
3-Boc-3,9-diazaspiro[5.5]undecane

Precaución: for Research Use Only

Formula: C14H26N2O2
MW: 254.3700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05861627
CAS Number: 173405-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR