tert-Butyl-3,8-diazabicyclo[3.2.1]octane-8-carboxylate 96%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3,8-diazabicyclo[3.2.1]octane-8-carboxylate, 3,8-Diazabicyclo[3.2.1]octane-8-carboxylic acid, 8-BOC protected

A136543-10G A136543-250MG A136543-25G A136543-5G A136543-1G A136543-100G
103802-318EA 17 USD
103802-318 77010-160 77010-162 77298-856 77298-858 77315-230 77683-205
tert-Butyl-3,8-diazabicyclo[3.2.1]octane-8-carboxylate 96%, Ambeed.Inc
tert-Butyl-3,8-diazabicyclo[3.2.1]octane-8-carboxylate

Precaución: For research use only.

Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
Boiling Pt: 295.4 °C (760 mm Hg)
Flash Pt: 132.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17016708
CAS Number: 149771-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR