tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-6-carboxylate 97%

Supplier: Ambeed
Warning

A105628-100MG A105628-10G A105628-25G A105628-1G A105628-5G A105628-250MG
103796-914EA 11 USD
103796-914 103822-650 76970-410 76970-412 76970-414 77309-518 77309-520 77592-122
tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-6-carboxylate 97%
tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₂
Boiling Pt: 276 °C (760 mm Hg)
Flash Pt: 121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17016736
CAS Number: 869494-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR