tert-Butyl 3,5-dioxopiperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A137688-100G A137688-10G A137688-25G A137688-5G A137688-1G
103802-486EA 6 USD
103802-486 76811-614 76811-616 76811-618 77298-918 77315-466
tert-Butyl 3,5-dioxopiperidine-1-carboxylate 97%
tert-Butyl 3,5-dioxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₄
MW: 213.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14706038
CAS Number: 396731-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR