tert-Butyl-3,5-dimethylpiperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-3,5-dimethylpiperazine

A132840-100MG A132840-250MG A132840-25G A132840-1G A132840-5G
103801-794EA 62 USD
103801-794 77010-014 77010-016 77010-018 77298-588 77314-380
tert-Butyl-3,5-dimethylpiperazine-1-carboxylate 97%
tert-Butyl-3,5-dimethylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Boiling Pt: 279.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07371499
CAS Number: 639068-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR