tert-Butyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A263046-250MG A263046-5G
103812-600EA 52 USD
103812-600 103852-496 76827-154 77304-864
tert-Butyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate 97%
tert-Butyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₇F₂NO₃
MW: 237.244 g/mol
Boiling Pt: 296.2±40.0 °C (760 mm Hg)
Flash Pt: 132.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16250751
CAS Number: 1209780-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR