tert-Butyl 3-(trifluoromethyl)-1,4,5,7-tetrahydro-6H-pyrazolo[3,4-c]pyridine-6-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(trifluoromethyl)-4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate

A270418-100MG A270418-250MG A270418-1G A270418-5G
103813-000EA 87 USD
103813-000 103852-904 76875-052 76875-054 77092-672 77092-674
tert-Butyl 3-(trifluoromethyl)-1,4,5,7-tetrahydro-6H-pyrazolo[3,4-c]pyridine-6-carboxylate 95%
tert-Butyl 3-(trifluoromethyl)-1,4,5,7-tetrahydro-6H-pyrazolo[3,4-c]pyridine-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₆F₃N₃O₂
MW: 291.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044974
CAS Number: 733757-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR