tert-Butyl 3-(tosyloxy)piperidine-1-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A102151-100MG
A102151-10G
A102151-250MG
103796-320EA
15
USD
103796-320
103822-014
77188-592
77188-594
77188-596
tert-Butyl 3-(tosyloxy)piperidine-1-carboxylate 95%
tert-Butyl 3-(tosyloxy)piperidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₇H₂₅NO₅S MW: 355.45 g/mol Boiling Pt: 475.3±38.0 °C (760 mm Hg) Flash Pt: 241.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07787194 CAS Number: 85275-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...