tert-Butyl 3-(bromomethyl)benzylcarbamate 98%

Supplier: Ambeed
Danger

A148542-1G A148542-250MG A148542-100MG
103803-940EA 145 USD
103803-940 103842-056 77317-520 77671-090 77883-176
tert-Butyl 3-(bromomethyl)benzylcarbamate 98%
tert-Butyl 3-(bromomethyl)benzylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD20257493
CAS Number: 220364-34-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR