1-Boc-3-(bromomethyl)azetidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 3-(bromomethyl)azetidine-1-carboxylate
A171240-100MG
A171240-250MG
A171240-25G
A171240-1G
A171240-5G
A171240-100G
103806-610EA
22
USD
103806-610
103844-884
76813-206
76813-208
76813-210
77082-024
77082-026
77671-430
1-Boc-3-(bromomethyl)azetidine 97%
1-Boc-3-(bromomethyl)azetidine
Precaución: for Research Use Only
Formula:
C₉H₁₆BrNO₂ MW: 250.136 g/mol Boiling Pt: 281.2±13.0 °C (760 mm Hg) Flash Pt: 123.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD16556174 CAS Number: 253176-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...