tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(aminomethyl)piperidine-1-carboxylate, tert-Butyl 3-(Aminomethyl)-1-piperidinecarboxylate, 3-(Aminomethyl)-1-Boc-piperidine, 3-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, 3-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester

A137906-1G A137906-25G A137906-5G
103802-528EA 12 USD
103802-528 77298-930 77298-932 77298-934
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate 97%
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Boiling Pt: 299.4 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01317792
CAS Number: 162167-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR