tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Tert-butyl 3-(aminomethyl)azetidine-1-carboxylate, 1-BOC-3-(aminomethyl)azetidine, tert-Butyl 3-(aminomethyl)azetidine-1-carboxylate, 3-Aminomethyl-azetidine-1-carboxylic acid tert-butyl ester

A356165-1G A356165-25G A356165-5G A356165-10G A356165-250MG
103826-160EA 9 USD
103826-160 103837-168 76840-302 76840-304 76840-306 77167-768 77167-770 77167-772
tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate 97%
tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Boiling Pt: 260.6 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861762
CAS Number: 325775-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR