3-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 3-(t-Butoxycarbonyl)phenylboronic acid pinacol ester, tert-Butyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

A309986-1G A309986-10G A309986-250MG A309986-25G
103814-608EA 8 USD
103814-608 103835-790 76838-564 77290-146 77290-148 77306-014
3-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester 97%
3-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₅BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD03453057
CAS Number: 903895-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR