1-Boc-5,6-dihydro-2H-pyridine-3-boronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Boc-5,6-Dihydro-2H-pyridine-3-boronic acid Pinacol ester, tert-Butyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate, 1-(tert-Butoxycarbonyl)-1,2,5,6-tetrahydro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, tert-Butyl 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydropyridine-1(2H)-carboxylate, 2-(1-Boc-1,2,5,6-tetrahydro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Boc-1,2,5,6-tetrahydro-3-pyridylboronic acid Pinacol ester, 2-[1-(tert-Butoxycarbonyl)-1,2,5,6-tetrahydro-3-pyridyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Boc-1,2,5,6-tetrahydro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 1-(tert-Butoxycarbonyl)-1,2,5,6-tetrahydro-3-pyridylboronic acid Pinacol ester, 1-(tert-Butoxycarbonyl)-5,6-Dihydro-2H-pyridine-3-boronic acid Pinacol ester
Precaución: for Research Use Only
Formula:
C₁₆H₂₈BNO₄ MW: 309.22 g/mol Boiling Pt: 348 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD10697911 CAS Number: 885693-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...