tert-Butyl 3-(2-hydroxyethyl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A211315-10G A211315-1G A211315-5G
103818-094EA 27 USD
103818-094 103848-788 77012-972 77087-192 77087-194
tert-Butyl 3-(2-hydroxyethyl)piperidine-1-carboxylate 97%
tert-Butyl 3-(2-hydroxyethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₃
MW: 229.316 g/mol
Boiling Pt: 324 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04114970
CAS Number: 146667-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR