Hydroxy-PEG1-t-butyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Hydroxy-PEG1-(CH2)2-Boc, tert-Butyl 3-(2-hydroxyethoxy)propanoate

A143357-250MG A143357-1G A143357-25G A143357-100G
103803-270EA 19 USD
103803-270 76984-292 77316-526 77316-528 77683-238
Hydroxy-PEG1-t-butyl ester 98%
Hydroxy-PEG1-t-butyl ester

Precaución: For research use only.

Formula: C₉H₁₈O₄
MW: 190.237 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22056310
CAS Number: 671802-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR