Amino-PEG1-t-Butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(2-aminoethoxy)propanoate

A312805-1G A312805-5G A312805-100MG A312805-250MG A312805-25G
103814-706EA 38 USD
103814-706 103835-896 76838-712 76838-714 77263-228 77263-230 77263-232
Amino-PEG1-t-Butyl ester 95%
Amino-PEG1-t-Butyl ester

Precaución: for Research Use Only

Formula: C₉H₁₉NO₃
MW: 189.252 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20727480
CAS Number: 1260092-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR