tert-Butyl 3-(1-aminoethyl)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A160975-100MG A160975-250MG A160975-1G
103805-436EA 267 USD
103805-436 77096-146 77096-148 77300-504
tert-Butyl 3-(1-aminoethyl)piperidine-1-carboxylate 95%
tert-Butyl 3-(1-aminoethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₂
MW: 228.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15209549
CAS Number: 1235439-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR