tert-Butyl-3-methylsulfonyloxypiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-((methylsulphonyl)oxy)piperidine-1-carboxylate

A110474-250MG A110474-1G A110474-10G
103797-668EA 13 USD
103797-668 103823-474 76718-070 77310-268 77666-618
tert-Butyl-3-methylsulfonyloxypiperidine-1-carboxylate 95%
tert-Butyl-3-methylsulfonyloxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₅S
MW: 279.36 g/mol
Boiling Pt: 407.2 °C (760 mm Hg)
Flash Pt: 200.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07787193
CAS Number: 129888-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR