tert-Butyl-2-oxospiro[indoline-3,3'-pyrrolidine]-1'-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-oxospiro[indoline-3,3'-pyrrolidine]-1'-carboxylate

A201457-1G A201457-5G A201457-250MG
103817-294EA 128 USD
103817-294 103847-934 77086-012 77086-014 77274-862
tert-Butyl-2-oxospiro[indoline-3,3'-pyrrolidine]-1'-carboxylate 95%
tert-Butyl-2-oxospiro[indoline-3,3'-pyrrolidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₃
MW: 288.3400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08458288
CAS Number: 205383-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR