1-(tert-Butoxycarbonyl)-2-pyrrolidinone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butoxycarbonyl)-2-oxopyrrolidine, tert-Butyl 2-oxopyrrolidine-1-carboxylate, Pyrrolidin-2-one, N-BOC protected

A118300-100G A118300-10G A118300-25G A118300-500G A118300-1G A118300-5G
103799-030EA 5 USD
103799-030 77189-486 77189-488 77189-490 77189-492 77311-706 77311-708
1-(tert-Butoxycarbonyl)-2-pyrrolidinone 95%
1-(tert-Butoxycarbonyl)-2-pyrrolidinone

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
Boiling Pt: 100…105 °C (0.5 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00674099
CAS Number: 85909-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR