tert-Butyl 2-oxo-1-oxa-3,8-diazaspiro[4.5]decane-8-carboxylate 95%

Supplier: Ambeed
Warning

A267999-100MG A267999-250MG A267999-1G A267999-5G
103812-890EA 95 USD
103812-890 103852-790 76787-132 76787-134 77092-544 77092-546
tert-Butyl 2-oxo-1-oxa-3,8-diazaspiro[4.5]decane-8-carboxylate 95%
tert-Butyl 2-oxo-1-oxa-3,8-diazaspiro[4.5]decane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₄
Melting Pt: 175…176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12913779
CAS Number: 169206-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR